siRNA Details
Gene Symbol: SIRPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130124.1 | - | 4 | 541 | ||
BM285386.1 | - | 3 | 267 | |||
BX537992.1 | - | 6 | 1296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
