siRNA Details
Gene Symbol: WFDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006103.3 | NP_006094.3 | 4 | 554 | ||
GenBank mRNA | AF330259.1 | - | 5 | 674 | ||
AF330260.1 | - | 3 | 399 | |||
AF330261.1 | - | 2 | 400 | |||
AK225528.1 | - | 4 | 553 | |||
BC039816.1 | - | 4 | 531 | |||
BC046106.1 | - | 4 | 530 | 5186932 | ||
BI759775.1 | - | 4 | 582 | |||
X63187.1 | - | 4 | 556 |
Gene Symbol: LRRC8D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC009486.1 | - | 1322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
