siRNA Details
Gene Symbol: WDR13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017883.4 | NP_060353.2 | 1 | 168 | ||
NR_029427.1 | 1 | 168 | ||||
GenBank mRNA | AF329819.4 | - | 1 | 168 | ||
AK314739.1 | - | 1 | 131 | |||
L08237.1 | - | 1 | 139 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
