siRNA Details
Gene Symbol: PCDH11X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168360.1 | NP_001161832.1 | 1 | 47 | ||
NM_001168361.1 | NP_001161833.1 | 1 | 47 | |||
NM_001168362.1 | NP_001161834.1 | 1 | 47 | |||
NM_001168363.1 | NP_001161835.1 | 1 | 47 | |||
NM_032968.4 | NP_116750.1 | 1 | 47 | |||
NM_032969.4 | NP_116751.1 | 1 | 47 | |||
XM_011530910.1 | XP_011529212.1 | 4 | 522 | |||
XM_011530911.2 | XP_011529213.1 | 6 | 810 | |||
XM_011530912.2 | XP_011529214.1 | 5 | 756 | |||
XM_011530913.2 | XP_011529215.1 | 5 | 756 | |||
XM_011530914.2 | XP_011529216.1 | 2 | 422 | |||
XM_011530915.2 | XP_011529217.1 | 5 | 756 | |||
XM_011530916.2 | XP_011529218.1 | 4 | 522 | |||
XM_017029416.1 | XP_016884905.1 | 1 | 31 | |||
XM_017029417.1 | XP_016884906.1 | 5 | 756 | |||
XM_017029418.1 | XP_016884907.1 | 5 | 756 | |||
XM_017029419.1 | XP_016884908.1 | 1 | 31 | |||
XM_017029420.1 | XP_016884909.1 | 5 | 756 | |||
XM_017029421.1 | XP_016884910.1 | 1 | 31 | |||
XM_017029422.1 | XP_016884911.1 | 2 | 422 | |||
GenBank mRNA | AB026187.1 | - | 1 | 848 | ||
AF332218.1 | - | 1 | 848 | |||
AF332219.1 | - | 1 | 848 | |||
AJ276804.1 | - | 4 | 592 | |||
AJ564931.1 | - | 1 | 47 | |||
AJ564932.1 | - | 1 | 848 | |||
AK299813.1 | - | 4 | 522 | |||
AK316263.1 | - | 4 | 522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
