siRNA Details
Gene Symbol: PCDH11X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011530911.2 | XP_011529213.1 | 3 | 572 | ||
XM_011530912.2 | XP_011529214.1 | 2 | 518 | |||
XM_011530913.2 | XP_011529215.1 | 2 | 518 | |||
XM_011530915.2 | XP_011529217.1 | 2 | 518 | |||
XM_017029417.1 | XP_016884906.1 | 2 | 518 | |||
XM_017029418.1 | XP_016884907.1 | 2 | 518 | |||
XM_017029420.1 | XP_016884909.1 | 2 | 518 |
Gene Symbol: PCDH11Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278619.1 | NP_001265548.1 | 2 | 523 | ||
XM_017030079.1 | XP_016885568.1 | 3 | 4753 | |||
XM_017030080.1 | XP_016885569.1 | 2 | 705 | |||
GenBank mRNA | AF277053.1 | - | 2 | 454 | ||
AJ564853.1 | - | 1 | 143 | |||
AJ564954.1 | - | 1 | 143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
