siRNA Details
Gene Symbol: CPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105533.1 | NP_001099003.1 | 6 | 1037 | ||
NM_024913.4 | NP_079189.4 | 7 | 1253 | |||
XM_017012649.1 | XP_016868138.1 | 6 | 1729 | |||
GenBank mRNA | AK025639.1 | - | 4 | 398 | ||
AK055624.1 | - | 6 | 1377 | |||
AK225313.1 | - | 4 | 398 | |||
AK289978.1 | - | 7 | 1250 | |||
AL832619.2 | - | 7 | 1208 | |||
AL832638.2 | - | 7 | 1077 | |||
AY358220.1 | - | 6 | 829 | |||
BC017764.1 | - | 3 | 250 | |||
BC018685.1 | - | 6 | 788 | |||
BC030538.2 | - | 3 | 267 | 3919732 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
