siRNA Details
Gene Symbol: POU6F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166018.1 | NP_001159490.1 | 7, 8 | 1169 | ||
NM_007252.3 | NP_009183.3 | 7, 8 | 1169 | |||
XM_011515113.2 | XP_011513415.1 | 6, 7 | 1189 | |||
GenBank mRNA | U91935.1 | - | 6, 7 | 1112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
