siRNA Details
Gene Symbol: AKNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317950.1 | NP_001304879.1 | 19 | 3891 | ||
NM_001317952.1 | NP_001304881.1 | 18 | 3449 | |||
NM_030767.5 | NP_110394.3 | 19 | 3836 | |||
XM_005252244.2 | XP_005252301.1 | 21 | 4008 | |||
XM_005252245.1 | XP_005252302.1 | 20 | 3866 | |||
XM_005252247.4 | XP_005252304.1 | 19 | 3887 | |||
XM_006717294.1 | XP_006717357.1 | 19 | 3820 | |||
XM_011519063.2 | XP_011517365.1 | 18 | 3499 | |||
XM_011519065.2 | XP_011517367.2 | 19 | 3479 | |||
XM_017015172.1 | XP_016870661.1 | 20 | 3606 | |||
GenBank mRNA | AF286341.1 | - | 9 | 1391 | ||
AK024431.1 | - | 17 | 2784 | |||
AK074040.1 | - | 18 | 2879 | |||
AK131082.2 | - | 19 | 3891 | |||
AK160382.1 | - | 19 | 3893 | |||
AL713755.1 | - | 2 | 2325 | |||
AL833220.1 | - | 16 | 2313 | |||
AY703039.1 | - | 20 | 3491 | |||
AY703040.1 | - | 18 | 3449 | |||
AY703043.1 | - | 18 | 3809 | |||
AY703044.1 | - | 17 | 3503 | |||
BC042202.1 | - | 5 | 566 | |||
BC055285.1 | - | 19 | 3836 | 6143155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
