siRNA Details
Gene Symbol: AKNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317950.1 | NP_001304879.1 | 13 | 3013 | ||
NM_001317952.1 | NP_001304881.1 | 12 | 2571 | |||
NM_030767.5 | NP_110394.3 | 13 | 2958 | |||
XM_005252244.2 | XP_005252301.1 | 15 | 3130 | |||
XM_005252245.1 | XP_005252302.1 | 14 | 2988 | |||
XM_005252247.4 | XP_005252304.1 | 13 | 3009 | |||
XM_005252248.2 | XP_005252305.1 | 13 | 3126 | |||
XM_006717294.1 | XP_006717357.1 | 13 | 2942 | |||
XM_011519063.2 | XP_011517365.1 | 12 | 2621 | |||
XM_011519064.2 | XP_011517366.1 | 13 | 3126 | |||
XM_011519065.2 | XP_011517367.2 | 13 | 2601 | |||
XM_017015172.1 | XP_016870661.1 | 14 | 2728 | |||
GenBank mRNA | AF286341.1 | - | 3 | 513 | ||
AK024431.1 | - | 11 | 1906 | |||
AK074040.1 | - | 12 | 2001 | |||
AK131082.2 | - | 13 | 3013 | |||
AK160382.1 | - | 13 | 3014 | |||
AL833220.1 | - | 10 | 1435 | |||
AY703039.1 | - | 14 | 2613 | |||
AY703040.1 | - | 12 | 2571 | |||
AY703043.1 | - | 12 | 2931 | |||
AY703044.1 | - | 11 | 2625 | |||
AY703045.1 | - | 4 | 708 | |||
BC055285.1 | - | 13 | 2958 | 6143155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
