siRNA Details
Gene Symbol: AKNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317950.1 | NP_001304879.1 | 3 | 1340 | ||
NM_001317952.1 | NP_001304881.1 | 2 | 898 | |||
NM_030767.5 | NP_110394.3 | 3 | 1285 | |||
XM_005252244.2 | XP_005252301.1 | 5 | 1457 | |||
XM_005252245.1 | XP_005252302.1 | 4 | 1315 | |||
XM_005252247.4 | XP_005252304.1 | 3 | 1336 | |||
XM_005252248.2 | XP_005252305.1 | 3 | 1453 | |||
XM_006717294.1 | XP_006717357.1 | 3 | 1269 | |||
XM_006717295.2 | XP_006717358.1 | 3 | 1453 | |||
XM_011519063.2 | XP_011517365.1 | 2 | 948 | |||
XM_011519064.2 | XP_011517366.1 | 3 | 1453 | |||
XM_011519065.2 | XP_011517367.2 | 3 | 928 | |||
XM_011519066.2 | XP_011517368.1 | 3 | 1453 | |||
XM_017015172.1 | XP_016870661.1 | 4 | 1055 | |||
GenBank mRNA | AB075848.1 | - | 3 | 1458 | ||
AK024431.1 | - | 1 | 233 | |||
AK074040.1 | - | 1 | 281 | |||
AK126136.1 | - | 3 | 1269 | |||
AK131082.2 | - | 3 | 1340 | |||
AK160382.1 | - | 3 | 1340 | |||
AY703039.1 | - | 4 | 940 | |||
AY703040.1 | - | 2 | 898 | |||
AY703041.1 | - | 4 | 654 | |||
AY703042.1 | - | 2 | 467 | |||
AY703043.1 | - | 2 | 1258 | |||
AY703044.1 | - | 1 | 952 | |||
BC055285.1 | - | 3 | 1285 | 6143155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
