siRNA Details
Gene Symbol: RLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006911.3 | NP_008842.1 | 2 | 503 | ||
XM_017015009.1 | XP_016870498.1 | 3 | 614 | |||
GenBank mRNA | A06846.1 | - | 2 | 376 | ||
A06926.1 | - | 2 | 376 | |||
A17329.1 | - | 2 | 376 | |||
BC005956.1 | - | 2 | 503 | 4296811 | ||
X00949.1 | - | 2 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
