siRNA Details
Gene Symbol: RAI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030665.3 | NP_109590.3 | 3 | 3294 | ||
XM_017024025.1 | XP_016879514.1 | 3 | 3391 | |||
XM_017024026.1 | XP_016879515.1 | 3 | 3697 | |||
XM_017024027.1 | XP_016879516.1 | 3 | 3633 | |||
XM_017024028.1 | XP_016879517.1 | 2 | 2980 | |||
GenBank mRNA | AB058723.1 | - | 3 | 3294 | ||
AJ271790.1 | - | 3 | 2684 | |||
AL834468.1 | - | 1 | 963 | |||
AY172136.1 | - | 3 | 3295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
