siRNA Details
Gene Symbol: HLA-DOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002119.3 | NP_002110.1 | 5 | 1709 | ||
GenBank mRNA | BC013183.1 | - | 5 | 1657 | 4340975 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
