siRNA Details
Gene Symbol: APBB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006051.3 | NP_006042.3 | 2 | 498 | ||
NM_133172.2 | NP_573418.2 | 2 | 498 | |||
NM_133173.2 | NP_573419.2 | 2 | 498 | |||
NM_133174.2 | NP_573420.2 | 2 | 498 | |||
GenBank mRNA | AB018247.1 | - | 2 | 190 | ||
AB049618.1 | - | 2 | 226 | |||
AB061224.1 | - | 2 | 139 | |||
AB079687.1 | - | 2 | 139 | |||
AB094990.1 | - | 2 | 139 | |||
AB209839.1 | - | 2 | 226 | |||
AF224708.1 | - | 2 | 159 | |||
AF224709.1 | - | 2 | 159 | |||
AF224710.1 | - | 2 | 159 | |||
AF224711.1 | - | 2 | 159 | |||
AK075280.1 | - | 2 | 461 | |||
AK125244.1 | - | 1 | 759 | |||
AK304824.1 | - | 2 | 265 | |||
AK307170.1 | - | 2 | 461 | |||
BC013158.1 | - | 2 | 498 | 3870148 | ||
BC125187.1 | - | 2 | 490 | 40122344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
