siRNA Details
Gene Symbol: UNC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133369.2 | NP_588610.2 | 2 | 475 | ||
XM_006714927.1 | XP_006714990.1 | 2 | 455 | |||
XM_006714928.1 | XP_006714991.1 | 2 | 455 | |||
XM_011534686.1 | XP_011532988.1 | 2 | 455 | |||
XM_011534687.2 | XP_011532989.1 | 2 | 455 | |||
GenBank mRNA | BC157824.1 | - | 2 | 358 | 9057043 | |
BI818609.1 | - | 2 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
