siRNA Details
Gene Symbol: UNC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133369.2 | NP_588610.2 | 3 | 502 | ||
XM_006714927.1 | XP_006714990.1 | 3 | 482 | |||
XM_006714928.1 | XP_006714991.1 | 3 | 482 | |||
XM_011534686.1 | XP_011532988.1 | 3 | 482 | |||
XM_011534687.2 | XP_011532989.1 | 3 | 482 | |||
GenBank mRNA | AK131380.1 | - | 4 | 379 | ||
BC033727.1 | - | 4 | 630 | |||
BC157824.1 | - | 3 | 385 | 9057043 | ||
BI818609.1 | - | 3 | 313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
