siRNA Details
Gene Symbol: TBCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134415.1 | NP_001127887.1 | 8 | 2640 | ||
NM_001286749.1 | NP_001273678.1 | 7 | 2261 | |||
NM_018138.4 | NP_060608.1 | 8 | 2291 | |||
GenBank mRNA | AK001422.1 | - | 8 | 2265 | ||
AK124362.1 | - | 7 | 2036 | |||
BC025748.1 | - | 8 | 2278 | 5200082 |
Gene Symbol: CRYGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC070241.1 | - | 1 | 212 | 30405628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
