siRNA Details
Gene Symbol: FOXP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012505.1 | NP_001012523.1 | 4 | 444 | ||
NM_001244808.1 | NP_001231737.1 | 4 | 547 | |||
NM_001244810.1 | NP_001231739.1 | 4 | 444 | |||
NM_001244816.1 | NP_001231745.1 | 3 | 349 | |||
NM_032682.5 | NP_116071.2 | 4 | 444 | |||
XM_005264737.4 | XP_005264794.1 | 4 | 453 | |||
XM_006713102.2 | XP_006713165.1 | 3 | 405 | |||
XM_006713104.2 | XP_006713167.1 | 2 | 375 | |||
XM_011533584.2 | XP_011531886.1 | 4 | 515 | |||
XM_011533585.2 | XP_011531887.1 | 3 | 367 | |||
XM_017006165.1 | XP_016861654.1 | 3 | 286 | |||
XM_017006168.1 | XP_016861657.1 | 4 | 453 | |||
GenBank mRNA | AF146696.1 | - | 2 | 181 | ||
AF250920.1 | - | 3 | 349 | |||
AK122710.1 | - | 4 | 547 | |||
BC005055.1 | - | 4 | 394 | |||
BC054815.1 | - | 4 | 394 | |||
BC071893.1 | - | 4 | 394 | |||
BC080521.1 | - | 4 | 394 | |||
BC131720.1 | - | 1 | 69 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
