siRNA Details
Gene Symbol: CENPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042426.1 | NP_001035891.1 | 4 | 611 | ||
NM_001809.3 | NP_001800.1 | 5 | 690 | |||
GenBank mRNA | BC000881.1 | - | 4 | 558 | 3461992 | |
BC002703.2 | - | 5 | 561 | 3626578 | ||
U14518.1 | - | 5 | 650 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
