siRNA Details
Gene Symbol: CENPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001809.3 | NP_001800.1 | 3 | 440 | ||
GenBank mRNA | BC002703.2 | - | 3 | 311 | 3626578 | |
BT007246.1 | - | 3 | 257 | |||
U14518.1 | - | 3 | 400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
