siRNA Details
Gene Symbol: CCDC88A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135597.1 | NP_001129069.1 | 16 | 3674 | ||
NM_001254943.1 | NP_001241872.1 | 16 | 3674 | |||
NM_018084.4 | NP_060554.3 | 16 | 3674 | |||
XM_005264418.4 | XP_005264475.1 | 16 | 3377 | |||
XM_005264421.2 | XP_005264478.1 | 16 | 3377 | |||
XM_005264426.2 | XP_005264483.1 | 16 | 3377 | |||
XM_011532965.2 | XP_011531267.1 | 16 | 3377 | |||
XM_011532966.2 | XP_011531268.1 | 16 | 3377 | |||
XM_011532967.2 | XP_011531269.1 | 16 | 3377 | |||
XM_011532968.2 | XP_011531270.1 | 16 | 3377 | |||
XM_017004476.1 | XP_016859965.1 | 16 | 3377 | |||
XM_017004477.1 | XP_016859966.1 | 16 | 3377 | |||
XM_017004478.1 | XP_016859967.1 | 16 | 3377 | |||
XM_017004479.1 | XP_016859968.1 | 16 | 3377 | |||
XM_017004480.1 | XP_016859969.1 | 14 | 3152 | |||
XM_017004481.1 | XP_016859970.1 | 16 | 3377 | |||
GenBank mRNA | AB125644.1 | - | 16 | 2832 | ||
AB201172.1 | - | 16 | 3366 | |||
BC056885.1 | - | 1 | 111 | |||
BC132736.1 | - | 16 | 3029 | 40146758 | ||
BC144320.1 | - | 16 | 3029 | 9052840 | ||
BU570532.1 | - | 2 | 611 | |||
BX537985.1 | - | 2 | 357 | |||
BX538154.1 | - | 2 | 283 | |||
BX648138.1 | - | 2 | 402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
