siRNA Details
Gene Symbol: NFE2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145412.3 | NP_001138884.1 | 5 | 2259 | ||
NM_001145413.3 | NP_001138885.1 | 5 | 2238 | |||
NM_001313900.1 | NP_001300829.1 | 5 | 2133 | |||
NM_001313901.1 | NP_001300830.1 | 5 | 2225 | |||
NM_001313902.1 | NP_001300831.1 | 4 | 2040 | |||
NM_001313903.1 | NP_001300832.1 | 5 | 1911 | |||
NM_001313904.1 | NP_001300833.1 | 5 | 2188 | |||
NM_006164.4 | NP_006155.2 | 5 | 2130 | |||
GenBank mRNA | AB208786.1 | - | 5 | 1641 | ||
AK297838.1 | - | 5 | 1634 | |||
AK304555.1 | - | 5 | 1679 | |||
AK314816.1 | - | 5 | 2131 | |||
AK316148.1 | - | 5 | 1582 | |||
BC011558.1 | - | 5 | 1688 | 4548874 | ||
BX649047.1 | - | 5 | 1669 | |||
BX649071.1 | - | 4 | 1470 | |||
HM446346.1 | - | 5 | 1582 | |||
S74017.1 | - | 5 | 1566 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
