siRNA Details
Gene Symbol: NFE2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145412.3 | NP_001138884.1 | 5 | 2602 | ||
NM_001145413.3 | NP_001138885.1 | 5 | 2581 | |||
NM_001313900.1 | NP_001300829.1 | 5 | 2476 | |||
NM_001313901.1 | NP_001300830.1 | 5 | 2568 | |||
NM_001313902.1 | NP_001300831.1 | 4 | 2383 | |||
NM_001313903.1 | NP_001300832.1 | 5 | 2254 | |||
NM_001313904.1 | NP_001300833.1 | 5 | 2531 | |||
NM_006164.4 | NP_006155.2 | 5 | 2473 | |||
GenBank mRNA | AB208786.1 | - | 5 | 1984 | ||
AK297838.1 | - | 5 | 1977 | |||
AK304555.1 | - | 5 | 2022 | |||
BC011558.1 | - | 5 | 2031 | 4548874 | ||
BX649047.1 | - | 5 | 2012 | |||
BX649071.1 | - | 4 | 1813 | |||
S74017.1 | - | 5 | 1909 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
