siRNA Details
Gene Symbol: TTLL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015072.4 | NP_055887.3 | 16, 17 | 1670 | ||
GenBank mRNA | AB023215.1 | - | 14, 15 | 1230 | ||
AK309748.1 | - | 17, 18 | 1652 | |||
AL136808.1 | - | 1, 2 | 79 | |||
AY237126.1 | - | 16, 17 | 1600 | |||
BC016625.1 | - | 3, 4 | 251 | |||
BC136472.1 | - | 16, 17 | 1560 | 9020461 | ||
BC136473.1 | - | 16, 17 | 1560 | 9020462 | ||
BX647530.1 | - | 9, 10 | 701 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
