siRNA Details
Gene Symbol: TTLL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015072.4 | NP_055887.3 | 22 | 2442 | ||
GenBank mRNA | AB023215.1 | - | 20 | 2002 | ||
AK309748.1 | - | 23 | 2424 | |||
AL136808.1 | - | 8 | 890 | |||
AY237126.1 | - | 22 | 2372 | |||
BC016625.1 | - | 9 | 1023 | |||
BC136472.1 | - | 22 | 2332 | 9020461 | ||
BC136473.1 | - | 22 | 2332 | 9020462 | ||
BX647530.1 | - | 15 | 1473 | |||
CR533554.1 | - | 7 | 820 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
