siRNA Details
Gene Symbol: TTLL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015072.4 | NP_055887.3 | 24 | 2751 | ||
GenBank mRNA | AB023215.1 | - | 22 | 2311 | ||
AK309748.1 | - | 25 | 2733 | |||
AL136808.1 | - | 10 | 1199 | |||
AY237126.1 | - | 24 | 2681 | |||
BC016625.1 | - | 11 | 1332 | |||
BC136472.1 | - | 24 | 2641 | 9020461 | ||
BC136473.1 | - | 24 | 2641 | 9020462 | ||
BX647530.1 | - | 17 | 1820 | |||
CR533554.1 | - | 9 | 1129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
