siRNA Details
Gene Symbol: TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003827.1 | NP_001003827.1 | 5, 6 | 923 | ||
NM_021616.5 | NP_067629.2 | 5, 6 | 927 | |||
GenBank mRNA | AB039902.1 | - | 5, 6 | 931 | ||
AF220143.1 | - | 5, 6 | 910 | |||
AF220144.1 | - | 6, 7 | 843 | |||
AK027876.1 | - | 5, 6 | 899 | |||
AL583914.1 | - | 4, 5 | 532 |
Gene Symbol: TRIM6-TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003819.3 | NP_001003819.1 | 11, 12 | 2091 | ||
GenBank mRNA | AB039903.1 | - | 11, 12 | 1999 | ||
BC136871.1 | - | 11, 12 | 2010 | 9020862 | ||
BC140722.1 | - | 11, 12 | 2010 | 9021590 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
