siRNA Details
Gene Symbol: PDZRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303139.1 | NP_001290068.1 | 7 | 1021 | ||
NM_001303140.1 | NP_001290069.1 | 7 | 884 | |||
NM_001303141.1 | NP_001290070.1 | 7 | 834 | |||
NM_001303142.1 | NP_001290071.1 | 7 | 1014 | |||
NM_015009.2 | NP_055824.1 | 9 | 1670 | |||
XM_017005942.1 | XP_016861431.1 | 8 | 1665 | |||
GenBank mRNA | AB029018.1 | - | 9 | 1670 | ||
AK074573.1 | - | 5 | 364 | |||
AK294761.1 | - | 7 | 825 | |||
AK299275.1 | - | 7 | 834 | |||
AK299560.1 | - | 7 | 868 | |||
AK307818.1 | - | 7 | 967 | |||
AK316244.1 | - | 7 | 834 | |||
AK316275.1 | - | 7 | 853 | |||
BC150623.1 | - | 9 | 1657 | 9056993 | ||
BC152417.1 | - | 9 | 1693 | 8862525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
