siRNA Details
Gene Symbol: PDZRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303139.1 | NP_001290068.1 | 4, 5 | 804 | ||
NM_001303140.1 | NP_001290069.1 | 4, 5 | 667 | |||
NM_001303141.1 | NP_001290070.1 | 4, 5 | 617 | |||
NM_001303142.1 | NP_001290071.1 | 4, 5 | 797 | |||
NM_015009.2 | NP_055824.1 | 6, 7 | 1453 | |||
XM_017005942.1 | XP_016861431.1 | 5, 6 | 1448 | |||
GenBank mRNA | AB029018.1 | - | 6, 7 | 1453 | ||
AK074573.1 | - | 2, 3 | 147 | |||
AK294761.1 | - | 4, 5 | 608 | |||
AK299275.1 | - | 4, 5 | 617 | |||
AK299560.1 | - | 4, 5 | 651 | |||
AK307818.1 | - | 4, 5 | 750 | |||
AK316244.1 | - | 4, 5 | 617 | |||
AK316275.1 | - | 4, 5 | 636 | |||
BC150623.1 | - | 6, 7 | 1440 | 9056993 | ||
BC152417.1 | - | 6, 7 | 1476 | 8862525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
