siRNA Details
Gene Symbol: DESI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297746.1 | NP_001284675.1 | 1 | 293 | ||
NM_016076.4 | NP_057160.2 | 1 | 293 | |||
NR_123735.1 | 1 | 293 | ||||
NR_123736.1 | 1 | 293 | ||||
XM_011544203.2 | XP_011542505.1 | 1 | 265 | |||
GenBank mRNA | AF151904.1 | - | 1 | 85 | ||
AF229834.1 | - | 1 | 34 | |||
AK314664.1 | - | 1 | 263 | |||
BC004485.2 | - | 1 | 240 | 3831754 | ||
BC020640.1 | - | 1 | 215 | |||
BI459676.1 | - | 1 | 297 | |||
CB854052.1 | - | 1 | 45 | |||
CN303012.1 | - | 1 | 204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
