siRNA Details
Gene Symbol: DESI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297746.1 | NP_001284675.1 | 3 | 408 | ||
NM_016076.4 | NP_057160.2 | 4 | 507 | |||
NR_123735.1 | 4 | 502 | ||||
NR_123736.1 | 3 | 413 | ||||
XM_011544203.2 | XP_011542505.1 | 5 | 592 | |||
XM_011544205.2 | XP_011542507.1 | 4 | 1158 | |||
XM_011544206.2 | XP_011542508.1 | 4 | 231 | |||
XM_011544207.2 | XP_011542509.1 | 3 | 1598 | |||
GenBank mRNA | AF151904.1 | - | 4 | 299 | ||
AF229834.1 | - | 3 | 149 | |||
AK314664.1 | - | 4 | 477 | |||
BC004485.2 | - | 4 | 454 | 3831754 | ||
BI459676.1 | - | 4 | 510 | |||
CB854052.1 | - | 3 | 165 | |||
CN303012.1 | - | 4 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
