siRNA Details
Gene Symbol: DESI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297746.1 | NP_001284675.1 | 3 | 431 | ||
NM_016076.4 | NP_057160.2 | 4 | 530 | |||
NR_123735.1 | 4 | 525 | ||||
NR_123736.1 | 3 | 436 | ||||
XM_011544203.2 | XP_011542505.1 | 5 | 615 | |||
XM_011544205.2 | XP_011542507.1 | 4 | 1181 | |||
XM_011544206.2 | XP_011542508.1 | 4 | 254 | |||
XM_011544207.2 | XP_011542509.1 | 3 | 1621 | |||
GenBank mRNA | AF151904.1 | - | 4 | 322 | ||
AF229834.1 | - | 3 | 172 | |||
AK314664.1 | - | 4 | 500 | |||
BC004485.2 | - | 4 | 477 | 3831754 | ||
CB854052.1 | - | 3 | 188 | |||
CN303012.1 | - | 4 | 436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
