siRNA Details
Gene Symbol: CSMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281956.1 | NP_001268885.1 | 22 | 3667 | ||
NM_052896.4 | NP_443128.2 | 22 | 3399 | |||
XM_017000185.1 | XP_016855674.1 | 22 | 3661 | |||
XM_017000186.1 | XP_016855675.1 | 22 | 3661 | |||
XM_017000187.1 | XP_016855676.1 | 22 | 3490 | |||
XM_017000188.1 | XP_016855677.1 | 22 | 3399 | |||
XM_017000189.1 | XP_016855678.1 | 22 | 3661 | |||
XM_017000190.1 | XP_016855679.1 | 16 | 2926 | |||
XM_017000191.1 | XP_016855680.1 | 22 | 3661 | |||
XM_017000192.1 | XP_016855681.1 | 22 | 3661 | |||
XM_017000193.1 | XP_016855682.1 | 22 | 3661 | |||
XM_017000194.1 | XP_016855683.1 | 22 | 3399 | |||
GenBank mRNA | AB212622.1 | - | 22 | 3667 | ||
AK127722.1 | - | 1 | 331 | |||
AK291019.1 | - | 1 | 172 | |||
AY210418.1 | - | 22 | 3399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
