siRNA Details
Gene Symbol: WWP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007013.3 | NP_008944.1 | 4 | 365 | ||
XM_005250760.3 | XP_005250817.1 | 3 | 326 | |||
XM_005250761.3 | XP_005250818.1 | 4 | 409 | |||
XM_011516802.2 | XP_011515104.1 | 3 | 126 | |||
XM_017012993.1 | XP_016868482.1 | 3 | 321 | |||
XM_017012994.1 | XP_016868483.1 | 3 | 316 | |||
XM_017012995.1 | XP_016868484.1 | 4 | 392 | |||
XM_017012996.1 | XP_016868485.1 | 3 | 310 | |||
XM_017012997.1 | XP_016868486.1 | 3 | 289 | |||
GenBank mRNA | AK292800.1 | - | 4 | 380 | ||
AL136739.1 | - | 2 | 95 | |||
AY043361.1 | - | 3 | 170 | |||
AY345857.1 | - | 4 | 399 | |||
BC036065.1 | - | 4 | 369 | 5296005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
