siRNA Details
Gene Symbol: WWP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007013.3 | NP_008944.1 | 14 | 1822 | ||
XM_005250760.3 | XP_005250817.1 | 13 | 1783 | |||
XM_005250761.3 | XP_005250818.1 | 14 | 1863 | |||
XM_011516802.2 | XP_011515104.1 | 13 | 1583 | |||
XM_017012993.1 | XP_016868482.1 | 13 | 1775 | |||
XM_017012994.1 | XP_016868483.1 | 13 | 1773 | |||
XM_017012995.1 | XP_016868484.1 | 12 | 1644 | |||
XM_017012996.1 | XP_016868485.1 | 11 | 1562 | |||
XM_017012997.1 | XP_016868486.1 | 11 | 1541 | |||
GenBank mRNA | AK292800.1 | - | 14 | 1837 | ||
AK310683.1 | - | 10 | 1196 | |||
AL136739.1 | - | 12 | 1552 | |||
AY043361.1 | - | 13 | 1627 | |||
AY345857.1 | - | 14 | 1856 | |||
BC036065.1 | - | 14 | 1826 | 5296005 | ||
U96113.1 | - | 7 | 976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
