siRNA Details
Gene Symbol: WWP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007013.3 | NP_008944.1 | 9 | 1200 | ||
XM_005250760.3 | XP_005250817.1 | 8 | 1161 | |||
XM_005250761.3 | XP_005250818.1 | 9 | 1241 | |||
XM_011516802.2 | XP_011515104.1 | 8 | 961 | |||
XM_017012993.1 | XP_016868482.1 | 8 | 1153 | |||
XM_017012994.1 | XP_016868483.1 | 8 | 1151 | |||
XM_017012995.1 | XP_016868484.1 | 7 | 1022 | |||
XM_017012996.1 | XP_016868485.1 | 6 | 940 | |||
XM_017012997.1 | XP_016868486.1 | 6 | 919 | |||
GenBank mRNA | AK292800.1 | - | 9 | 1215 | ||
AK310683.1 | - | 5 | 574 | |||
AL136739.1 | - | 7 | 930 | |||
AY043361.1 | - | 8 | 1005 | |||
AY345857.1 | - | 9 | 1234 | |||
BC036065.1 | - | 9 | 1204 | 5296005 | ||
U96113.1 | - | 2 | 354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
