siRNA Details
Gene Symbol: OBSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173408.1 | NP_001166879.1 | 6 | 2502 | ||
NM_001173431.1 | NP_001166902.1 | 6 | 2502 | |||
NM_015311.2 | NP_056126.1 | 6 | 2502 | |||
XM_005246424.4 | XP_005246481.1 | 6 | 2526 | |||
XM_005246427.4 | XP_005246484.1 | 6 | 2517 | |||
XM_011510857.2 | XP_011509159.1 | 6 | 2529 | |||
XM_011510863.2 | XP_011509165.1 | 6 | 2521 | |||
XM_011510864.2 | XP_011509166.1 | 6 | 2521 | |||
XM_011510865.2 | XP_011509167.1 | 6 | 2521 | |||
XM_011510866.2 | XP_011509168.1 | 6 | 2520 | |||
XM_017003696.1 | XP_016859185.1 | 6 | 2528 | |||
XM_017003697.1 | XP_016859186.1 | 6 | 2528 | |||
XM_017003698.1 | XP_016859187.1 | 6 | 2527 | |||
XM_017003699.1 | XP_016859188.1 | 6 | 2528 | |||
XM_017003700.1 | XP_016859189.1 | 6 | 2526 | |||
GenBank mRNA | AB014557.2 | - | 6 | 1697 | ||
AF035292.1 | - | 3 | 628 | |||
AK025946.1 | - | 5 | 955 | |||
BC007201.1 | - | 6 | 2247 | |||
EF063637.1 | - | 6 | 2248 | |||
EF063638.1 | - | 6 | 2245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
