siRNA Details
Gene Symbol: FAM210B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080821.2 | NP_543011.2 | 3 | 1087 | ||
GenBank mRNA | AJ311123.1 | - | 3 | 1048 | ||
AK075565.1 | - | 3 | 1034 | |||
BC017725.1 | - | 3 | 1044 | 3887324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
