siRNA Details
Gene Symbol: PPP4R1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003505.3 | 9 | 846 | |||
GenBank mRNA | AK057583.1 | - | 9 | 846 | ||
AK299330.1 | - | 9 | 983 | |||
AL833409.1 | - | 6 | 1809 | |||
BC100979.1 | - | 10 | 989 | |||
BC100980.1 | - | 10 | 989 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
