siRNA Details
Gene Symbol: PRICKLE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013397.5 | NP_037529.3 | 2 | 169 | ||
GenBank mRNA | BC110458.1 | - | 1 | 96 | ||
BC110459.1 | - | 1 | 96 | 40024767 | ||
DA875485.1 | - | 2 | 146 | |||
DA957684.1 | - | 2 | 169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
