siRNA Details
Gene Symbol: HIST1H3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003530.4 | NP_003521.2 | 1 | 206 | ||
GenBank mRNA | AK316611.1 | - | 1 | 206 | ||
BC031333.1 | - | 1 | 125 | 3608479 | ||
BQ051491.1 | - | 1 | 149 | |||
DB242999.1 | - | 1 | 206 |
Gene Symbol: HIST1H2AD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021065.3 | NP_066409.1 | 1 | 206 | ||
GenBank mRNA | AK312127.1 | - | 1 | 206 | ||
BC093807.1 | - | 1 | 195 | 7939652 | ||
BC093809.1 | - | 1 | 198 | 7939654 | ||
BC128035.1 | - | 1 | 156 | 40021702 | ||
CR541970.1 | - | 1 | 156 | |||
CR541999.1 | - | 1 | 156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
