siRNA Details
Gene Symbol: HIST1H3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003530.4 | NP_003521.2 | 1, 2 | 441 | ||
GenBank mRNA | AK316611.1 | - | 1, 2 | 441 | ||
BQ051491.1 | - | 1, 2 | 384 | |||
DB242999.1 | - | 1, 2 | 441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
