siRNA Details
Gene Symbol: ATXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000332.3 | NP_000323.2 | 1 | 46 | ||
NM_001128164.1 | NP_001121636.1 | 1 | 46 | |||
GenBank mRNA | AK311274.1 | - | 1 | 17 | ||
AK311649.1 | - | 1 | 16 | |||
BC011026.1 | - | 1 | 49 | |||
BC047894.1 | - | 1 | 32 | |||
BC063120.1 | - | 1 | 27 | |||
BF670717.1 | - | 46 | ||||
BP365416.1 | - | 1 | 17 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
