siRNA Details
Gene Symbol: ATXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000332.3 | NP_000323.2 | 3 | 343 | ||
NM_001128164.1 | NP_001121636.1 | 2 | 294 | |||
GenBank mRNA | AK311274.1 | - | 2 | 265 | ||
BC014548.1 | - | 2 | 251 | |||
BC039236.1 | - | 2 | 174 | |||
BC047894.1 | - | 2 | 280 | |||
BC063120.1 | - | 2 | 275 | |||
BC092446.1 | - | 1 | 62 | |||
BC113082.1 | - | 1 | 95 | |||
BP365416.1 | - | 2 | 265 | |||
X79204.1 | - | 3 | 307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
