siRNA Details
Gene Symbol: ATXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000332.3 | NP_000323.2 | 6 | 633 | ||
NM_001128164.1 | NP_001121636.1 | 5 | 584 | |||
GenBank mRNA | AK311274.1 | - | 5 | 555 | ||
BC029401.1 | - | 4 | 394 | |||
BP365416.1 | - | 5 | 555 | |||
X79204.1 | - | 6 | 597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
