siRNA Details
Gene Symbol: BEST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282613.1 | NP_001269542.1 | 7 | 945 | ||
NM_001282614.1 | NP_001269543.1 | 9 | 1207 | |||
NM_032735.2 | NP_116124.2 | 9 | 1207 | |||
NM_152439.3 | NP_689652.2 | 5 | 554 | |||
XM_006719256.3 | XP_006719319.1 | 8 | 995 | |||
XM_011537961.1 | XP_011536263.1 | 7 | 815 | |||
XM_011537962.2 | XP_011536264.1 | 6 | 578 | |||
XM_011537963.1 | XP_011536265.1 | 6 | 586 | |||
XM_011537965.2 | XP_011536267.1 | 3 | 325 | |||
GenBank mRNA | AF440758.1 | - | 9 | 1152 | ||
AK096459.1 | - | 9 | 1208 | |||
AY515706.1 | - | 8 | 980 | |||
BC151138.1 | - | 7 | 814 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
