siRNA Details
Gene Symbol: BEST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282613.1 | NP_001269542.1 | 4 | 638 | ||
NM_001282614.1 | NP_001269543.1 | 6 | 900 | |||
NM_032735.2 | NP_116124.2 | 6 | 900 | |||
NM_152439.3 | NP_689652.2 | 3 | 400 | |||
XM_006719256.3 | XP_006719319.1 | 5 | 688 | |||
XM_011537961.1 | XP_011536263.1 | 4 | 508 | |||
XM_011537962.2 | XP_011536264.1 | 3 | 271 | |||
XM_011537963.1 | XP_011536265.1 | 3 | 279 | |||
GenBank mRNA | AF440758.1 | - | 6 | 845 | ||
AK096459.1 | - | 6 | 901 | |||
AY515706.1 | - | 5 | 673 | |||
BC151138.1 | - | 4 | 507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
