siRNA Details
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020945.1 | NP_065996.1 | 38 | 6577 | ||
XM_005270004.3 | XP_005270061.1 | 38 | 6692 | |||
XM_011539986.2 | XP_011538288.2 | 38 | 6695 | |||
XM_011539987.2 | XP_011538289.2 | 38 | 6698 | |||
XM_011539988.2 | XP_011538290.1 | 39 | 6776 | |||
XM_011539990.2 | XP_011538292.1 | 38 | 6723 | |||
XM_017016463.1 | XP_016871952.1 | 39 | 6619 | |||
XM_017016464.1 | XP_016871953.1 | 20 | 3006 | |||
GenBank mRNA | AB046827.1 | - | 5 | 760 | ||
AK074085.1 | - | 18 | 2635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
