siRNA Details
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020945.1 | NP_065996.1 | 54, 55 | 8519 | ||
XM_005270004.3 | XP_005270061.1 | 54, 55 | 8634 | |||
XM_011539986.2 | XP_011538288.2 | 54, 55 | 8637 | |||
XM_011539987.2 | XP_011538289.2 | 54, 55 | 8640 | |||
XM_011539988.2 | XP_011538290.1 | 55, 56 | 8718 | |||
XM_017016463.1 | XP_016871952.1 | 55, 56 | 8561 | |||
XM_017016464.1 | XP_016871953.1 | 36, 37 | 4948 | |||
GenBank mRNA | AB046827.1 | - | 22, 23 | 2702 | ||
AK024502.1 | - | 5, 6 | 686 | |||
AK074085.1 | - | 35, 36 | 4577 | |||
AK074443.1 | - | 17, 18 | 1922 | |||
AK301736.1 | - | 8, 9 | 975 | |||
AK310374.1 | - | 8, 9 | 1019 | |||
AL833669.1 | - | 15, 16 | 1648 | |||
BC047574.1 | - | 15, 16 | 1588 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
