siRNA Details
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020945.1 | NP_065996.1 | 62 | 9712 | ||
XM_005270004.3 | XP_005270061.1 | 62 | 9827 | |||
XM_011539986.2 | XP_011538288.2 | 62 | 9830 | |||
XM_011539987.2 | XP_011538289.2 | 62 | 9812 | |||
XM_011539988.2 | XP_011538290.1 | 63 | 9911 | |||
XM_017016463.1 | XP_016871952.1 | 63 | 9754 | |||
XM_017016464.1 | XP_016871953.1 | 44 | 6141 | |||
GenBank mRNA | AB046827.1 | - | 30 | 3895 | ||
AK024502.1 | - | 12 | 4269 | |||
AK074085.1 | - | 42 | 5559 | |||
AK074443.1 | - | 25 | 3114 | |||
AL833669.1 | - | 23 | 2841 | |||
BC015694.2 | - | 7 | 1043 | |||
BC032420.1 | - | 7 | 1131 | |||
BC047574.1 | - | 23 | 2781 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
